Learn More
Witeg™ Biogen™ Triple Baffled Erlenmeyer Flask with Rimmed Neck
Product Code. 10440713
Product Code. 10440713 Supplier Witeg™ Supplier No. 5 507 205
Provide improved agitation and gas transfer in these triple-baffled flasks. Witeg™ Biogen™ Triple Baffled Erlenmeyer Flask with Rimmed Neck is designed with three internal indents for greater mixing and long, straight necks that accommodate metal caps and reduce splashing.
  • Graduated.
  • Labelling space.
  • Triple baffled.
  • Rimmed neck
  • Long neck.
Specifications
Capacity (Metric) Media 2,000 mL
Description Triple-baffled
Diameter (Metric) Center Neck 50 mm
For Use With (Equipment) Orbital shaker
Height (Metric) 280 mm
Neck Style Long, rimmed
Shape Erlenmeyer
Writing Surface Yes
Graduated Yes
Closure Type Cap