Learn More
Fisherbrand™ Biological Microscope for Epifluorescence
The five position fluorescent turret adapter contains four combined sets of excitation and barrier filters.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
- Compound and trinocular
- Fifth neutral position allows for standard brightfield viewing
- External power supply powers the external lamphouse (HB0-100W mercury bulb)
- Blue light filter is suitable for FITC fluorescent antibody observation, green light filter is suitable for TRITC fluorescent antibody observation
- Adjustable condensing lens, iris diaphragm, anti-UV protection screen and a slide plate with two positions, closed and open for fluorescence light transmission
- With fluorescence free oil, two spare fuses, instruction manual and blocking plate for stage
Specifications
Brightfield Abbe - NA 0.90 | |
WF10X/20 mm | |
High Pressure Mercury Bulb | |
Magnum T/Epi Fluor. | |
Fluorescence | |
Quintuple reversed and revolving with click stop | |
220/240 V | |
50/60 Hz |
Iris | |
Trinocular, Optical, with Vertical Photo/Video Port | |
4X, 10X, 40X, 100X | |
Centrifuge Rotor | |
Blue and green filter, 1 spare bulb, 2 spare fuses, immersion oil 10mL, soft rubber eyecups, dust cover and instruction manual. | |
22 kg | |
220/240 V |