Learn More
Beckman Coulter Life Sciences Biomek i3 Cell Transfection Bundle
Description
- Saving precious space without compromising performance and versatility
- 15 ANSI/SLAS standard pipettable and 5 flex positions
- Dimensions: 73.2(W) x 78.5(D) x 82.6 cm(H)
- Gripper supports labware up to 12.8 cm in height and 725 g in weight, and it's capable of stacking up to four plates
- Ready for On-deck and Off-deck integration (Left, Right, Back and Down)
- Getting results you can trust with accurate flexible pipetting
- Pipetting Volume Range: 1 μL to 1000 μL with air displacement technology
- Fixed-8 Pipetting Pod allows selective loading of 1to 8 tips in any sequence
- Uses standard Biomek i-Series 96-format disposable tips
- Lab automation made simple for any skill level
- Beckman Connect enables secure remote diagnostics and trouble shooting
- Guided Labware setup
- Method Options Selector Step Builder
- Camera-enabled error log
- Comfort and safe operation
- Installed UV-C light
- Positive or negative pressure HEPA-filtration (optional)
- Deck Observation System (2 cameras)
- Safety shields
Specifications
Specifications
| Product Type | Automated Liquid Handler |
| Channels | 8 |
| Material | Aluminum, Stainless Steel, EPDM, Polyethylene |
| Volume (Metric) | 1 to 1000 μL |
Products not intended or validated for use in the diagnosis of disease or other conditions.