Learn More
Biopterin-d3, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Biopterin-d3 |
| CAS | 1217838-71-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 2H3 H8 N5 O3 |
| Quantity | 0.5 mg |
| Synonym | rel-2-Amino-6-[(1R,2S)-1,2-dihydroxypropyl-3,3,3-d3]-4(3H)-pteridinone, DL-erythro-Biopterin-3',3',3'-D3, DL-erythro-Biopterin-d3, Biopterin-d3 |
| InChI Formula | InChI=1S/C9H11N5O3/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7/h2-3,6,15-16H,1H3,(H3,10,11,13,14,17)/t3-,6-/m0/s1/i1D3 |
| SMILES | [2H]C([2H])([2H])[C@H](O)[C@H](O)c1cnc2nc(N)nc(O)c2n1 |
| IUPAC Name | (1R,2S)-1-(2-amino-4-hydroxypteridin-6-yl)-3,3,3-trideuteriopropane-1,2-diol |
| Molecular Weight (g/mol) | 240.23 |
| Show More |