Learn More
Tocris Bioscience™ Biotin-11-dUTP
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
Description
Biotin-11-dUTP is a biotin-labeled dUTP. Commonly used in PCR, nick translation, and reverse transcription. Also frequently used in apoptosis assays. Biotin-labeled DNA may be detected with streptavidin-HRP conjugates.
Specifications
Specifications
| Chemical Name or Material | 2 '-Deoxy-5-[(1 E)-3-[[6-[[5-[(3 AS,4 S,6 aR)-hexahydro-2-oxo-1 H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]-1-oxohexyl]amino]-1-propen-1-yl]uridine 5 '-(tetra(triethylammonium) triphosphate) |
| Quantity | 200 μL |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C28H45N6O17P3S0.4(C2H5)3N |
| Purity | 95% |