missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin Antibody (1D4-C5), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23510R
This item is not returnable.
View return policy
Description
Biotin Monoclonal antibody specifically detects Biotin in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA
Specifications
| Biotin | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Flow Cytometry, ELISA | |
| 1D4-C5 | |
| Flow Cytometry, ELISA | |
| Vitamin B4 | |
| Biotinylated Limulus hemocyanin | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction