missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Biotin Antibody (rBTN/8819) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27043AF700
This item is not returnable.
View return policy
Description
Biotin Monoclonal antibody specifically detects Biotin in Nonspecies specific samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),In-situ Hybridization,Immunocytochemistry
Specifications
| Biotin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Vitamin B4 | |
| Biotinylated sheep immunoglobulin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Immunocytochemistry | |
| rBTN/8819 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?