missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated Human IgG Heavy chain Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech Biotin-67760-20UL
This item is not returnable.
View return policy
Description
The Human IgG Heavy chain antibody from Proteintech is a mouse monoclonal antibody generated with nature protein of Human IgG Heavy chain. This antibody recognizes human antigen. The Human IgG Heavy chain antibody has been validated for the following applications: WB analysis. This antibody is biotin.Specifications
| IgG Heavy chain | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| Human | |
| ∼50 kDa | |
| Liquid |
| Western Blot | |
| 1C6B4 | |
| Biotin | |
| nature protein | |
| 20 μL | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction