Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (Hyb-8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34646AF750
Product Code. 18130983
  • € 563.00 / 0.10mL
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal specifically detects Biotin (Vitamin B7 or Vitamin H) in All Species samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biotin (Vitamin B7 or Vitamin H)
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Mouse
Protein A or G purified
Primary
All species
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Hyb-8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H
Biotinylated sheep immunoglobulin
0.1 mL
It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This monoclonal antibody is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.
Store at 4°C in the dark.