missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotinidase/BTD Antibody (12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06622MFV500
This item is not returnable.
View return policy
Description
Biotinidase/BTD Monoclonal antibody specifically detects Biotinidase/BTD in Human samples. It is validated for ELISA
Specifications
| Biotinidase/BTD | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 12 | |
| ELISA | |
| biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Biotinidase/BTD (Uniprot#: P43251; Met1-Asp543). | |
| 0.1 mL | |
| Signal Transduction | |
| 686 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction