missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotinidase/BTD Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97093AF750
This item is not returnable.
View return policy
Description
Biotinidase/BTD Polyclonal specifically detects Biotinidase/BTD in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Biotinidase/BTD | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
| Produced in rabbits immunized with purified, recombinant Biotinidase/BTD (P43251; Met1-Asp543). | |
| 0.1 mL | |
| Signal Transduction | |
| 686 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction