Learn More
(+)-Biotinyl 3,6,9-Trioxaundecanediamine, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | (+)-Biotinyl 3,6,9-Trioxaundecanediamine |
| CAS | 359860-27-8 |
| Molecular Formula | C18H34N4O5S |
| Quantity | 25 mg |
| Synonym | (3aS,4S,6aR)-N-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, EZ-Link Amine-PEO3-Biotin, EZ-Link Biotin-PEO-LC Amine |
| InChI Formula | InChI=1S/C18H34N4O5S/c19-5-7-25-9-11-27-12-10-26-8-6-20-16(23)4-2-1-3-15-17-14(13-28-15)21-18(24)22-17/h14-15,17H,1-13,19H2,(H,20,23)(H2,21,22,24)/t14-,15-,17-/m0/s1 |
| SMILES | NCCOCCOCCOCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12 |
| IUPAC Name | 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethyl]pentanamide |
| Molecular Weight (g/mol) | 418.55 |