Learn More
Thermo Scientific™ Biotinyl-Phe-Ala-Asp fluoromethyl ketone
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 5 mg |
| Description | Off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | A cell permeable, biotinylated analog of the cathespin inhibitor Z-Phe-Ala-Fluoromethyl Ketone (Z-FA-FMK). Z-FA-FMK inhibits the cysteine proteases, Cathepsins B and L,1 |
| Molecular Formula | C27H36FN5O7S |
| Formula Weight | 593.67 |
| Synonym | Biotin-FAD-FMK; Biotin-Phe-Ala-Asp-FMK |
| Solubility | Soluble in DMSO or DMF. |
| Product Type | Biotinyl-Phe-Ala-Asp fluoromethyl ketone |
| Form | Powder |