Learn More
Cytiva BioTrace™ NT Nitrocellulose Transfer Membrane
Product Code. 17124571
missing translation for 'orderingAttributeHoverText'
Dimensions (LxW):
20 x 20 cm
30 cm x 3 m
7 x 8.5 cm
missing translation for 'unitSize'
1 roll
Pack of 10
This item is not returnable. Rückgaberichtlinie anzeigen
Product Code. 17124571
missing translation for 'mfr': Cytiva 66485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BioTrace NT is a pure unsupported nitrocellulose membrane that has high binding capacity for proteins and nucleic acids.
  • High tensile strength makes it less likely to tear or crack than other brands of nitrocellulose
  • High binding capacity for both proteins and nucleic acids
  • Ideal for colony/plaque lifts and protein transfers
  • Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
TRUSTED_SUSTAINABILITY
Specifications
Packaging Type Roll
Description Non-sterile
Material BioTrace NT
Pore Size 0.2 μm
Dimensions (LxW) 30 cm x 3 m
Thickness 101.6 to 190.5 μm