missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIRC3 Monoclonal Antibody (OTI2G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800039
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.Specifications
| BIRC3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13489 | |
| BIRC3 | |
| Human recombit protein fragment corresponding to amino acids 255-529 of human BIRC3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2G7 | |
| Unconjugated | |
| BIRC3 | |
| AIP1, API2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49, c-IAP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 330 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction