Learn More
BIRC3 Monoclonal Antibody (OTI2G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800039S
Encompass_Preferred
 View more versions of this product
Product Code. 17547412
  • € 242.00 / 30µL
Description
BIRC3 Monoclonal antibody specifically detects BIRC3 in Human samples. It is validated for Western Blot
Specifications
Specifications
BIRC3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75317
Mouse
Affinity chromatography
RUO
219333
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2G7
Unconjugated
USP12
AIP 2; AIP1; Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW107670; baculoviral IAP repeat containing 3; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 3; Birc2; Birc3; Cellular inhibitor of apoptosis 2; cIAP1; cIAP-1; cIAP2; C-IAP2; cIAP-2; H HIAP1; H MALT2; HAIP1; HIAP1; hIAP-1; HIAP2; IAP homolog; IAP homolog C; IAP1; IAP-1; IAP2; Inhibitor of apoptosis protein 1; MALT2; mammalian IAP homolog C; MIAP1; mIAP-1; MIAP2; MIHB; MIHC; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; RNF49; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Human recombinant protein fragment corresponding to amino acids 61-251 of human USP12 produced in E.coli.