missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIRC5/Survivin Monoclonal Antibody (OTI3G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | BIRC5/Survivin |
|---|---|
| Clone | OTI3G3 |
| Concentration | 0.73 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17516361
|
Origene Technologies
TA502236S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17596351
|
Origene Technologies
TA502236 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BIRC5/Survivin Monoclonal antibody specifically detects BIRC5/Survivin in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| BIRC5/Survivin | |
| 0.73 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 332 | |
| Primary |
| OTI3G3 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O15392 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title