missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birch pollen Bet V1 Antibody (BV16), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81093MFV610
This item is not returnable.
View return policy
Description
Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA
Specifications
| Birch pollen Bet V1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Plant | |
| Purified |
| Western Blot, ELISA | |
| BV16 | |
| Western Blot, ELISA | |
| Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
| Birch pollen Bet V1-A, a major allergen of the European white birch. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction