missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-di-tert-butylphenyl) Hydrogen Phosphate, TRC

Product Code. 30138618
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138618 500 mg 500mg
30133731 250 mg 250mg
17993755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138618 Supplier Toronto Research Chemicals Supplier No. TRCB178380500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69284-93-1
Molecular Formula C28 H43 O4 P
Molecular Weight (g/mol) 474.61
Synonym Bis(2,4-di-tert-butylphenyl) Phosphate, Phenol, 2,4-bis(1,1-dimethylethyl)-, 1,1'-(hydrogen phosphate), Phenol, 2,4-bis(1,1-dimethylethyl)-, hydrogen phosphate (9CI), Bis(2,4-di-tert-butylphenyl) phosphate, Bis(2,4-di-tert-butylphenyl) Phosphate;
IUPAC Name bis(2,4-ditert-butylphenyl) hydrogen phosphate
SMILES CC(C)(C)c1ccc(OP(=O)(O)Oc2ccc(cc2C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C

Specifications

Analyte or Component Names Bis(2,4-di-tert-butylphenyl) Hydrogen Phosphate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C28H43O4P/c1-25(2,3)19-13-15-23(21(17-19)27(7,8)9)31-33(29,30)32-24-16-14-20(26(4,5)6)18-22(24)28(10,11)12/h13-18H,1-12H3,(H,29,30)
Solution Type Neat
Industry Type Industrial QC
Quantity 500 mg
Formula Weight 474.2899
Percent Purity >95%
Chemical Name or Material Bis(2,4-di-tert-butylphenyl) Hydrogen Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.