Learn More
Bis(2-chloroethyl) (2-Chloroethyl)phosphonate (>80%), TRC
Product Code. 30342468
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342468 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342468 Supplier Toronto Research Chemicals Supplier No. TRCB825605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(2-chloroethyl) (2-Chloroethyl)phosphonate
CAS 6294-34-4
Molecular Formula C6H12Cl3O3P
Quantity 100 mg
Synonym Bis(2-chloroethyl) P-(2-chloroethyl)phosphonate, 2-Chloroethanol (2-chloroethyl)phosphonate (2:1), Bis(2-chloroethyl) (2-chloroethyl)phosphonate, Bis(beta-chloroethyl) beta-chloroethylphosphonate, Bis-chloroethyl 2-chloroethanephosphonate
InChI Formula InChI=1S/C6H12Cl3O3P/c7-1-4-11-13(10,6-3-9)12-5-2-8/h1-6H2
SMILES ClCCOP(=O)(CCCl)OCCCl
IUPAC Name 1-chloro-2-[2-chloroethoxy(2-chloroethyl)phosphoryl]oxyethane
Molecular Weight (g/mol) 269.49
Formula Weight 267.959