Learn More
Bis(2-chloroethyl)methylamine Hydrochloride, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Mechlorethamine Hydrochloride |
| CAS | 55-86-7 |
| Molecular Formula | C5 H11 Cl2 N . Cl H |
| Quantity | 250 mg |
| Synonym | Mechlorethamine Hydrochloride, 2-Chloro-N-(2-chloroethyl)-N-methylethanamine hydrochloride, 2,2'-Dichloro-N-methyldiethylamine hydrochloride, N-Methyl-2,2'-dichlorodiethylamine hydrochloride, Di(chloroethyl)methylamine hydrochloride, Methylbis(β-chloroethyl)amine hydrochloride, Methyldi(2-chloroethyl)amine hydrochloride, Chlormethine hydrochloride, Caryolysine hydrochloride, Cloramin hydrochloride, Dichloren hydrochloride, Embichen hydrochloride, Embikhine hydrochloride, Erasol hydrochloride, Mustargen hydrochloride hydrochloride, Mustine hydrochloride hydrochloride, Nitrogranulogen |
| InChI Formula | InChI=1S/C5H11Cl2N.ClH/c1-8(4-2-6)5-3-7;/h2-5H2,1H3;1H |
| SMILES | Cl.CN(CCCl)CCCl |
| IUPAC Name | 2-chloro-N-(2-chloroethyl)-N-methylethanamine;hydrochloride |
| Molecular Weight (g/mol) | 192.51 |
| Formula Weight | 191.0035 |