Learn More
Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite, TRC
Product Code. 30347890
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347890 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347890 Supplier Toronto Research Chemicals Supplier No. TRCB4200005G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite
CAS 102690-88-0
Molecular Formula C12 H22 N3 O2 P
Quantity 5 g
InChI Formula InChI=1S/C12H22N3O2P/c1-11(2)15(12(3)4)18(16-9-5-7-13)17-10-6-8-14/h11-12H,5-6,9-10H2,1-4H3
SMILES CC(C)N(C(C)C)P(OCCC#N)OCCC#N
IUPAC Name 3-[2-cyanoethoxy-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile
Molecular Weight (g/mol) 271.3
Formula Weight 271.145