Learn More
Bis(4-chlorophenyl) disulfide, 98+%
Product Code. 11481627
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481627 5 g 5g
11491627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11481627 Supplier Thermo Scientific Alfa Aesar Supplier No. B25604.06
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
CAS: 1142-19-4 | C12H8Cl2S2 | 287.216 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1142-19-4
Molecular Formula C12H8Cl2S2
Molecular Weight (g/mol) 287.216
MDL Number MFCD00013642
InChI Key ZIXXRXGPBFMPFD-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl disulfide, bis 4-chlorophenyl disulfide, disulfide, bis 4-chlorophenyl, ddds, ddds pesticide, disulfide, bis p-chlorophenyl, p-chlorophenyl disulfide, 4-chlorophenyl disulfide, bis p-chlorophenyl disulfide, 4,4'-dichloro diphenyl disulfide
PubChem CID 14360
IUPAC Name 1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1SSC2=CC=C(C=C2)Cl)Cl
Specifications
Melting Point 71°C to 74°C
Quantity 5 g
Beilstein 1644663
Formula Weight 287.23
Percent Purity ≥98%
Chemical Name or Material Bis(4-chlorophenyl) disulfide