Learn More
Bis(8-methyl-1-nonyl) Phthalate-3,4,5,6-d4, CDN
Specifications
Specifications
| Chemical Name or Material | Diisodecyl phthalate-d4 |
| CAS | 1346604-79-2 |
| Molecular Formula | C28 2H4 H42 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-bis(8-methylnonyl) ester, Diisodecyl Phthalate-d4, Phthalic Acid Di-isodecyl Ester (3,4,5,6)-D4 |
| InChI Formula | InChI=1 S/C28H46O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,19-20,23-24 H,5-12,15-18,21-22H2,1-4H3/i13D,14 D,19 D,20 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCC(C)C)c(C(=O)OCCCCCCCC(C)C)c1[2 H] |
| IUPAC Name | bis(8-methylnonyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 450.69 |
| Formula Weight | 450.3647 g/mol |
| Show More |