Learn More
Bis(8-methylnonyl) Cyclohex-4-ene-1,2-dicarboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(8-methylnonyl) Cyclohex-4-ene-1,2-dicarboxylate |
| CAS | 87826-26-4 |
| Molecular Formula | C28H50O4 |
| Quantity | 500 mg |
| Synonym | Bis(8-methylnonyl) Cyclohex-4-ene-1,2-dicarboxylate |
| InChI Formula | InChI=1S/C28H50O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,23-26H,5-12,15-22H2,1-4H3 |
| SMILES | O=C(C1C(C(OCCCCCCCC(C)C)=O)CC=CC1)OCCCCCCCC(C)C |
| IUPAC Name | bis(8-methylnonyl) cyclohex-4-ene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 450.69 |
| Formula Weight | 450.37 |