missing translation for 'onlineSavingsMsg'
Learn More

Bis(diethylamino)phenylphosphine, Thermo Scientific Chemicals

Catalog Number p-7112895
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1636-14-2
C14H25N2P
252.34
MFCD00137954
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine, bis diethylamino phenylphosphine, diethylamino phenyl phosphanyl diethylamine, acmc-20almr, tetraethylphenylphosphonous diamide, phenylphosphonous tetraethyldiamide, bis diethylamino phenylphosphine 97, n-diethylamino phenyl phosphanyl-n-ethylethanamine, phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(N(CC)CC)C1=CC=CC=C1
This item is not returnable. View return policy
1636-14-2
C14H25N2P
252.34
MFCD00137954
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine, bis diethylamino phenylphosphine, diethylamino phenyl phosphanyl diethylamine, acmc-20almr, tetraethylphenylphosphonous diamide, phenylphosphonous tetraethyldiamide, bis diethylamino phenylphosphine 97, n-diethylamino phenyl phosphanyl-n-ethylethanamine, phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(N(CC)CC)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9710g/mL
Flash Point 106°C
Linear Formula C6H5P(N(CH2CH3)2)2
Quantity 5 g
Specific Gravity 0.971
Formula Weight 252.34
Chemical Name or Material Bis(diethylamino)phenylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'