Learn More
Bis(tributylammonium) Pyrophosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(tributylammonium) Pyrophosphate |
| CAS | 5975-18-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 2 C12 H27 N . H4 O7 P2 |
| Quantity | 1 g |
| Synonym | Diphosphoric acid, compd. with N,N-dibutyl-1-butanamine (1:2), Tributylamine, pyrophosphate (2:1) (8CI), Tributylamine, pyrophosphate (7CI), 1-Butanamine, N,N-dibutyl-, (diphosphate) (2:1) (9CI), Pyrophosphoric acid, compd. with tributylamine (1:2) (8CI), Bis(tri-n-butylammonium) pyrophosphate, Bis(tributylammonium) pyrophosphate |
| InChI Formula | InChI=1S/2C12H27N.H4O7P2/c2*1-4-7-10-13(11-8-5-2)12-9-6-3;1-8(2,3)7-9(4,5)6/h2*4-12H2,1-3H3;(H2,1,2,3)(H2,4,5,6) |
| SMILES | CCCCN(CCCC)CCCC.CCCCN(CCCC)CCCC.OP(=O)(O)OP(=O)(O)O |
| IUPAC Name | N,N-dibutylbutan-1-amine;phosphono dihydrogen phosphate |
| Molecular Weight (g/mol) | 548.67 |
| Show More |