Learn More
Bis(trimethylsilyl)methane, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2117-28-4 |
| Molecular Formula | C7H20Si2 |
| MDL Number | MFCD00008275 |
| Quantity | 1 g |
| SMILES | C[Si](C[Si](C)(C)C)(C)C |
| Molecular Weight (g/mol) | 160.40 |
| Percent Purity | 98% |
| Chemical Name or Material | Bis(trimethylsilyl)methane |