Learn More
Thermo Scientific Chemicals BIS-TRIS, 0.2M buffer soln., pH 7.4
Product Code. p-7057398
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. 15464619
Brand: Thermo Scientific Chemicals J62209.AE
Request Bulk or Custom Format
CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO
Specifications
Color Colorless
Concentration 0.2 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Chemical Name or Material BIS-TRIS