missing translation for 'onlineSavingsMsg'
Learn More

Bis(vinylsulfonylmethyl) Ether, TRC

Product Code. 30334868
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334868 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334868 Supplier Toronto Research Chemicals Supplier No. TRCB563898250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(vinylsulfonylmethyl) Ether
CAS 26750-50-5
Molecular Formula C6H10O5S2
Quantity 250 mg
InChI Formula InChI=1S/C6H10O5S2/c1-3-12(7,8)5-11-6-13(9,10)4-2/h3-4H,1-2,5-6H2
SMILES C=CS(=O)(=O)COCS(=O)(=O)C=C
IUPAC Name 1-(ethenylsulfonylmethoxymethylsulfonyl)ethene
Molecular Weight (g/mol) 226.27
Formula Weight 225.997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.