missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Standard Solution (Spectro ECON), Chem Lab™

Product Code. 11989022
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11989022

Brand: Chem-Lab CL01.0225.0500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 1000 μg/mL
Quantity 500 mL
compliance-icons
Product Identifier
  • Bismuth standard solution 1000 µg/ml
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.