missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisoprolol hemifumarate, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Description
Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.
Specifications
Specifications
| Chemical Name or Material | Bisoprolol hemifumarate |
| CAS | 104344-23-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H31NO4.1/2C4H4O4 |
| Quantity | 200 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (65.19 mM) ∣H2O : 20 mg/mL (26.08 mM; Need ultrasonic) |
| SMILES | OC(CNC(C)C)COC1=CC=C(COCCOC(C)C)C=C1.OC(/C=C/C(O)=O)=O.[1/2] |
| Molecular Weight (g/mol) | 766.96 |
| Formula Weight | 766.96 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction