Learn More
Bisoprolol hemifumarate, MedChemExpress
Product Code. 16468076
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16468076
Brand: Medchem Express HYB0076
MedChemExpress Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.
Bisoprolol hemifumarate is a potent, selective and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. Bisoprolol hemifumarate has the potential for hypertension, coronary artery disease and stable ventricular dysfunction research.
Specifications
Chemical Name or Material Bisoprolol hemifumarate
CAS 104344-23-2
Color White
Purity Grade Notes Research
Molecular Formula C18H31NO4.1/2C4H4O4
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (65.19 mM) ∣H2O : 20 mg/mL (26.08 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=C(COCCOC(C)C)C=C1.OC(/C=C/C(O)=O)=O.[1/2]
Molecular Weight (g/mol) 766.96
Formula Weight 766.96
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only