missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A Bis(diphenyl phosphate), TRC

Product Code. 30134707
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134707 2.5 g 2.5g
30003261 1 g 1g
30006840 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134707 Supplier Toronto Research Chemicals Supplier No. TRCB5195302.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5945-33-5
Molecular Formula C39 H34 O8 P2
Molecular Weight (g/mol) 692.63
Synonym Phosphoric acid, (1-methylethylidene)di-4,1-phenylene tetraphenyl ester (9CI), Phosphoric acid, diphenyl ester, diester with 4,4'-isopropylidenediphenol (7CI), Phosphoric acid, isopropylidenedi-p-phenylene tetraphenyl ester (8CI), 2,2-Bis[4-[bis(phenoxy)phosphoryloxy]phenyl]propane, 4,4'-(Isopropylidenediphenyl) bis(diphenyl phosphate), AD 191, ADK Stab FP 600, ADK Stab FP 700, BADP, BDP, BDP 30LH, BPA-DP, Bisphenol A bis(diphenyl phosphate), Bisphenol A tetraphenyl diphosphate, CG 963, CR 41, CR 7415, CR 741S, CR 742, E 890, FP 600, FP 700, FP 750, Fyrolflex BDP, NcendX P 30, PhireGuard BDP, Tetraphenyl bisphenol A bisphosphate, WSFR-BDP, WSFR-BDP-N 2
IUPAC Name [4-[2-(4-diphenoxyphosphoryloxyphenyl)propan-2-yl]phenyl] diphenyl phosphate
SMILES CC(C)(c1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)cc1)c4ccc(OP(=O)(Oc5ccccc5)Oc6ccccc6)cc4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C39H34O8P2/c1-39(2,31-23-27-37(28-24-31)46-48(40,42-33-15-7-3-8-16-33)43-34-17-9-4-10-18-34)32-25-29-38(30-26-32)47-49(41,44-35-19-11-5-12-20-35)45-36-21-13-6-14-22-36/h3-30H,1-2H3
Formula Weight 692.1729
Chemical Name or Material Bisphenol A Bis(diphenyl phosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.