missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A diglycidyl ether resin

Product Code. p-7046536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343696 25 g 25g
11353696 100 g 100g
11363696 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343696 Supplier Thermo Scientific Alfa Aesar Supplier No. L14817.14
Add to basket
This item is not returnable. View return policy

CAS: 1675-54-3 | C21H24O4 | 340.419 g/mol

Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1675-54-3
Molecular Formula C21H24O4
Molecular Weight (g/mol) 340.419
MDL Number MFCD00080480
InChI Key LCFVJGUPQDGYKZ-UHFFFAOYSA-N
Synonym bisphenol a diglycidyl ether, 2,2-bis 4-glycidyloxyphenyl propane, epoxide a, epophen el 5, badge, dian diglycidyl ether, diglycidyl bisphenol a, epi-rez 510, bpdge, dgeba
PubChem CID 2286
ChEBI CHEBI:34578
IUPAC Name 2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
SMILES CC(C)(C1=CC=C(C=C1)OCC2CO2)C3=CC=C(C=C3)OCC4CO4

Specifications

Density 1.17
Refractive Index 1.573
Quantity 25 g
Beilstein 299026
Solubility Information Soluble in 100% ethanol,dimethyl sulfoxide (100 mM),dimethyl formamide,chloroform,methanol,and ethanol (50 mM). Insoluble in water.
Formula Weight 340.42
Chemical Name or Material Bisphenol A diglycidyl ether
compliance-icons compliance-icons
Product Identifier
  • Bisphenol A diglycidyl ether
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bisphenol A diglycidyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.