missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol E-13C6, TRC

Product Code. 30012313
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012313 5 mg 5mg
30011333 10 mg 10mg
30004910 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012313 Supplier Toronto Research Chemicals Supplier No. TRCB5194825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C813C6H14O2
Molecular Weight (g/mol) 220.22
Synonym 4,4'-Ethylidenedi-phenol, 1,1-Bis(4-hydroxyphenyl)ethane, 4,4'-Ethylidenebisphenol, 4,4'-Ethylidenediphenol, Bisphenol AD
IUPAC Name 4-[1-(4-hydroxyphenyl)ethyl](1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-ol
SMILES O[13C]([13C]([H])=[13C]1[H])=[13C]([H])[13C]([H])=[13C]1C(C)C2=CC=C(O)C=C2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H14O2/c1-10(11-2-6-13(15)7-3-11)12-4-8-14(16)9-5-12/h2-10,15-16H,1H3/i2+1,3+1,6+1,7+1,11+1,13+1
Percent Purity >95%
Chemical Name or Material Bisphenol E-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.