Learn More
Tocris Bioscience™ BIX 01294
GREENER_CHOICE
Product Code. 18045612
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18045612
Brand: Tocris Bioscience™ 3364/50
GLP and G9a inhibitor
GLP and G9a histone lysine methyltransferase inhibitor (IC50 values are 0.7 and 1.7 μM respectively) that displays no activity at other histone methyltransferases up to 37 μM. Modulates H3K9me2 levels in mammalian cells and potentiates induction of pluripotent stem cells from somatic cells in vitro. Also inhibits H3K36 methylation by oncoproteins NSD1, NSD2 and NSD3 (IC50 values are 40 - 112 μM).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1392399-03-9
Molecular Formula C28H41Cl3N6O2
Specifications
Quantity 50 mg
Formula Weight 600.02
Purity >98%
Molecular Weight (g/mol) 600.03
Chemical Name or Material BIX 01294