Description
- High density halogen free ribbons
- Excellent solvent resistance
- High heat and scratch resistance
- Compatible with Brady™ BBP12 and BBP11 label printers
- Resin ribbon formulation
Specifications
Specifications
| Color | Black |
| Material | Resin |
| Length (Metric) | 70 m |
| Width (Metric) | 65 mm |
| Alternate Part Number | R6000HF-65X70/O |