Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Brand:  Axygen™ HTF-1000-CBK-HTR-5
Product Code. 17302623
  • € 662.00 / Pack of 3840
Estimated Shipment: 07-10-2026
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Non-sterile
Not Autoclavable
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Liquid Level Sensing, Automation
Hamilton™ MICROLAB™ Nimbus
Filtered
Black
DNase, RNase and Pyrogen free
Hanging Tip Rack
5 Racks/Pk, 8 Pks/Cs.