missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLAME/SLAMF8 Antibody (242), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90657AF405
This item is not returnable.
View return policy
Description
BLAME/SLAMF8 Monoclonal specifically detects BLAME/SLAMF8 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BLAME/SLAMF8 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56833 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 242 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B Lymphocyte Activator Macrophage Expressed, BCM-Like Membrane Protein, B-Lymphocyte Activator Macrophage Expressed, CD353, CD353 Antigen, SBBI42, SLAM Family Member 8, SLAMF8 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BLAME/SLAMF8 (NP_083360.2; Met1-Asp231). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction