missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLAP75 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92213-0.1ml
This item is not returnable.
View return policy
Description
BLAP75 Polyclonal antibody specifically detects BLAP75 in Human, Mouse samples. It is validated for Western Blot
Specifications
| BLAP75 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| 75 kDa, BLM-associated protein 75 kDa, BLM-associated protein of 75 kDa, chromosome 9 open reading frame 76, FAAP75, FLJ12888, homolog of yeast RecQ-mediated genome instability 1 (RMI1), recQ-mediated genome instability protein 1, RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae), RMI2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human RMI1 (NP_079221.2). MNVTSIALRAETWLLAAWHVKVPPMWLEACINWIQEENNNVNLSQAQMNKQVFEQWLLTDLRDLEHPLLPDGILEIPKGELNGFYALQINSLVDVSQPAYSQIQKLRGKNTTNDLVTAEAQVTPKPWEAKPSRMLMLQLTDGIVQIQGMEYQPIPILHSDLPPGTKILIYGNISFRLGVLLLKPENVKVLGGEVDALLEEYAQEKVLARLIGEPDLVVSVIPNNSNENIPRVTDVLDPALGPSDEELLASLDENDELTANNDTSSERCFTTGSSSNTIPTRQSSFEPEFVISPRPKEEPSNLSIHVMDGELDDFSLEEALLLEETVQKEQMETKELQPLTFNRNADRSIE | |
| 0.1 mL | |
| Direct Reversal of DNA Damage | |
| 80010 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction