Learn More
Brand™ Silberbrand™ Borosilicate Glass Class B Volumetric Flask
Product Code. 10211741 Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Limit of Error:
±0.06 mL
±0.09 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
±0.9 mL
±1.8 mL
Unit Size:
Each
Pack of 2
Product Code. 10211741
Brand: Brand™ 36753
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Silberbrand™ Borosilicate Glass Class B Volumetric Flask includes polypropylene stopper. White enamel DE-M marking provides quick volume check.
  • Manufactured from Borosilicate Glass
  • Complies with DIN EN ISO 1042
  • Class B
  • Calibrated to contain (TC, In.)
  • Includes polypropylene stopper
  • White enamel DE-M marking provides quick volume check
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042
Closure Material Polypropylene
Color Clear
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Limit of Error ±0.6 mL
No. per Case 1 Piece
Tolerance ±0.6 mL
Class B
Closure Size 24/29