Learn More
Brand™ Blaubrand™ Eterna™ Class AS Bulb Pipets, with Certificate
Shop All Brand Products
Click to view available options
:
Capacity: 10 mL
Description
- Manufactured from AR-GLAS™ Soda-lime glass.
- DIN EN ISO 648. Calibrated to deliver (TD, Ex).
- Waiting time 5 seconds.
- Delivered with batch number and one batch certificate per packing unit of the manufacturer.
- Delivered with individual conformity certificate.
Specifications
Specifications
| Calibration | To Deliver |
| Certifications/Compliance | DIN EN ISO 648, DIN 12691 |
| Class | AS |
| Material | AR-GLAS, Soda Lime Glass |
| Graduated | Yes |
| Graduation Color | Amber |
| Graduation Type | DE-M Marking |
| Tolerance | 0.02 mL |
| Color | Red |
| Volume (Metric) | 10 mL |
| Show More |