Learn More
BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, with Certificate
Product Code. 15422423 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
200 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 15422423
Brand: BRAND™ 956980
Only null left
Add to basket
Add to basket
Indispensable for preparing dilutions and standard solutions. Delivered with individual USP conformity certificate.
  • Includes with glass stopper with dripping tip
  • Contrast blue enamel print on transparent flasks ensures optimum combination of resistance and readability
  • Available in all ISO sizes from 5 to 2000mL
  • Available in transparent and amber colors
  • Made of low expansion coefficient, chemically resistant borosilicate 3.3 glass
  • Delivered with individual USP conformity certificate
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP
Color Transparent
Capacity (Metric) 200 mL
Material Glass
Class A
Closure Size NS 14/23