missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Blk Human Protein, Active

Product Code. 17300284
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17300284

Brand: Eurofins DiscoverX 14517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, human, N-terminal His6 tagged, recombinant full-length human Blk, for use in Kinase Assays.

Activation of B cells by various ligands is accompanied by activation of src-family protein tyrosine kinases. Cross-linking of the B-cell receptor leads to the activation of src-family PTKs BLK (B-cell lymphocyte kinase), Fyn, and Lyn. Specific B and T cell progenitor subsets are preferentially susceptible to transformation by BLK and suggest a role for BLK in the control of proliferation during B cell development. BLK protein tyrosine kinase is found primarily in hematopoietic cells and inhibitors may be of therapeutic use in the treatment of B cell lymphoma.

GenBank NM_001715. The recombinant protein contains one residue which is in conflict with NM_001715. This is M287V and is reported inGenBank S76617 and BC007371.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 58.8 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥64%
Protein Blk, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.