missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112369
This item is not returnable.
View return policy
Description
BLK Polyclonal Antibody for Western Blot, IHC (P), ELISA
B-lymphoid tyrosine kinase (BLK) is a non-receptor tyrosine kinase that belongs to the Src family of kinases. BLK plays an important role in B-cell activation. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
Specifications
| BLK | |
| Polyclonal | |
| Unconjugated | |
| BLK | |
| B lymphocyte kinase; B lymphoid kinase; B lymphoid tyrosine kinase; Blk; BLK nonreceptor tyrosine kinase; BLK proto-oncogene, Src family tyrosine kinase; MGC10442; MODY11; p55-Blk; tyrosine-protein kinase Blk | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12143, 640 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16277, P51451 | |
| BLK | |
| Recombinant Human Tyrosine-protein kinase Blk protein (1-280AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction