missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLMH/Bleomycin Hydrolase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | BLMH/Bleomycin Hydrolase |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682498
|
Novus Biologicals
NBP2-99780-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661849
|
Novus Biologicals
NBP2-99780-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| BLMH/Bleomycin Hydrolase | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 0.2 um filtered solution in PBS | |
| 642 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
| Produced in rabbits immunized with purified, recombinant Human BLMH/Bleomycin Hydrolase (Accession#: NP_000377.1; Ser 2-Glu 455). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title