missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-542
This item is not returnable.
View return policy
Description
BLNK Polyclonal specifically detects BLNK in Mouse samples. It is validated for Western Blot.
Specifications
| BLNK | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) with 15mM Sodium Azide | |
| BLNK | |
| A fusion protein representing amino acids 171-356 of human BLNK. | |
| Protein A purified | |
| RUO | |
| Primary | |
| The polyclonal antibody reacts with mouse SLP65 BLNK, a cytosolic adaptor protein identified as two phosphoproteins migrating at 68 and 70 kDa in SDS/PAGE (alternatively spliced forms of human SLP65). | |
| Store at 4C. Do not freeze. |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:1000 | |
| AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| Rabbit | |
| 70 kDa | |
| 0.1 mg | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 29760 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction