Learn More
BLNK Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808522S
Encompass_Preferred
 View more versions of this product
Product Code. 17579853
  • € 242.00 / 30µL
Description
BLNK Monoclonal antibody specifically detects BLNK in Human samples. It is validated for Western Blot
Specifications
Specifications
BLNK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6UX39
Mouse
Affinity chromatography
RUO
401138
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2B2
Unconjugated
AMTN
AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa
Full length human recombinant protein of human AMTN produced in E.coli.
30 μL