missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLNK Monoclonal Antibody (OTI4C9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527007
This item is not returnable.
View return policy
Description
BLNK Monoclonal Antibody for Western Blot, IHC (P)
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
Specifications
| BLNK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WV28 | |
| BLNK | |
| Human recombinant protein fragment corresponding to amino acids 1-292 of BLNK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C9 | |
| Unconjugated | |
| BLNK | |
| AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction